Learn More
Biomatik Corporation Phospho-TIF-IA (Ser649) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51688
Encompass_Preferred
Phospho-TIF-IA (Ser649) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TIF-IA (Ser649) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9NYV6. Gene ID: 54700. Target Synonyms: RNA polymerase I-specific transcription initiation factor RRN3; RRN3; transcription initiation factor IA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1796