Learn More
Biomatik Corporation Phospho-NMDAR2B (Tyr1336) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51660
Encompass_Preferred
Phospho-NMDAR2B (Tyr1336) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NMDAR2B (Tyr1336) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13224. Gene ID: 2904. Target Synonyms: Glutamate [NMDA] receptor subunit epsilon 2 precursor; GRIN2B; hNR3; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3; NMDE2; NME2; NR2B; NR3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1768