Learn More
Biomatik Corporation Phospho-AIRE (Ser156) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51664
Encompass_Preferred
Phospho-AIRE (Ser156) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AIRE (Ser156) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43918. Gene ID: 326. Target Synonyms: AIRE1; APECED; APS1; APSI; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy; PGA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1772