Learn More
Biomatik Corporation Phospho-Claudin 3 (Tyr219) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51669
Encompass_Preferred
Phospho-Claudin 3 (Tyr219) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Claudin 3 (Tyr219) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15551. Gene ID: 1365. Target Synonyms: Claudin-3; Clostridium perfringens enterotoxin receptor 2; CPE-receptor 2; CPE-R 2; Ventral prostate.1 protein homolog; HRVP1; CLDN3; C7orf1; CPETR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1777