Learn More
Biomatik Corporation Phospho-Epo-R (Tyr368) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51296
Encompass_Preferred
Phospho-Epo-R (Tyr368) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Epo-R (Tyr368) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P19235. Gene ID: 2057. Target Synonyms: EPO-R; EPOR; Erythropoietin receptor precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9040