Learn More
Biomatik Corporation Phospho-Abl (Tyr412) Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA51704
Encompass_Preferred
Phospho-Abl (Tyr412) Colorimetric Cell-Based ELISA for detecting Phospho-Abl (Tyr412) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42684. Gene ID: 25; 27. Target Synonyms: ABL1; JTK7; Proto-oncogene tyrosine-protein kinase ABL1; c-ABL; p150. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1808