Learn More
Biomatik Corporation Phospho-TERF1 (Ser219) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51383
Encompass_Preferred
Phospho-TERF1 (Ser219) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TERF1 (Ser219) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P54274. Gene ID: 7013. Target Synonyms: NIMA-interacting protein 2; PIN2; TERF1; TRF; TTAGGG repeat binding factor 1; Telomeric protein Pin2/TRF1; Telomeric repeat binding factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1541