Learn More
Biomatik Corporation Phospho-Bcr (Tyr177) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51408
Encompass_Preferred
Phospho-Bcr (Tyr177) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Bcr (Tyr177) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11274. Gene ID: 613. Target Synonyms: BCR; BCR protein; BCR1; Breakpoint cluster region protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1563