Learn More
Biomatik Corporation Phospho-eNOS (Thr494) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51441
Encompass_Preferred
Phospho-eNOS (Thr494) Colorimetric Cell-Based ELISA Kit for detecting Phospho-eNOS (Thr494) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29474. Gene ID: 4846. Target Synonyms: cNOS; Constitutive NOS; EC-NOS; ECNOS; Endothelial NOS; Nitric-oxide synthase; endothelial; NOS; type III; NOS3; NOSIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1585