Learn More
Biomatik Corporation Phospho-FAK (Tyr925) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51451
Encompass_Preferred
Phospho-FAK (Tyr925) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FAK (Tyr925) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05397. Gene ID: 5747. Target Synonyms: FADK 1; FAK1; Focal adhesion kinase 1; pp125FAK; Protein- tyrosine kinase 2; PTK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1594