Learn More
Biomatik Corporation Phospho-PI3-kinase p85-α/γ (Tyr467/199) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51380
Encompass_Preferred
Phospho-PI3-kinase p85-α/γ (Tyr467/199) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PI3-kinase p85-α/γ (Tyr467/199) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27986; Q92569. Gene ID: 5295. Target Synonyms: GRB1; P85A; PI3-kinase p85-alpha subunit; PI3K; PI3K p85-alpha; Phosphatidylinositol 3-kinase regulatory alpha subunit; PtdIns-3-kinase p85-alpha; P55G; PI3-kinase p85-gamma subunit; PI3K p85-gamma; Phosphatidylinositol 3-kinase regulatory gamma subunit; PtdIns-3-kinase p85-gamma; p55PIK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9049