Learn More
Biomatik Corporation Phospho-Merlin (Ser518) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51506
Encompass_Preferred
Phospho-Merlin (Ser518) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Merlin (Ser518) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35240. Gene ID: 4771. Target Synonyms: MERL; moesin-ezrin-radizin-like protein; neurofibromatosis type 2 gene; neurofibromatosis-2; Neurofibromin 2; NF-2; NF2; SCH; Schwannomerlin; schwannomerlin; Schwannomin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1643