Learn More
Biomatik Corporation Phospho-MKK3 (Ser189) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51509
Encompass_Preferred
Phospho-MKK3 (Ser189) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MKK3 (Ser189) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P46734. Gene ID: 5606. Target Synonyms: dual specificity mitogen-activated protein kinase kinase 3; kinase MKK3; MAP kinase kinase 3; MAP2K3; MAPK/ERK kinase 3; MAPKK 3; MEK3; MP2K3; PRKMK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1645