Learn More
Biomatik Corporation Phospho-p56 Dok-2 (Tyr299) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51430
Encompass_Preferred
Phospho-p56 Dok-2 (Tyr299) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p56 Dok-2 (Tyr299) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60496. Gene ID: 9046. Target Synonyms: Docking protein 2; Dok-R; Dok-related protein; DOK2; DokR; downstream of tyrosine kinase 2; FRIP; IL-4 receptor interacting protein; p56(dok-2). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9059