Learn More
Biomatik Corporation Phospho-p27 Kip1 (Ser10) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51528
Encompass_Preferred
Phospho-p27 Kip1 (Ser10) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p27 Kip1 (Ser10) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P46527. Gene ID: 1027. Target Synonyms: CDKN1B; CDN1B; Cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor p27; KIP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1662