Learn More
Biomatik Corporation Phospho-Retinoblastoma (Ser807) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51555
Encompass_Preferred
Phospho-Retinoblastoma (Ser807) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Retinoblastoma (Ser807) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06400. Gene ID: 5925. Target Synonyms: P105-RB; PP105; PP110; RB-1; RB1; retinoblastoma gene; Retinoblastoma-associated protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1685