Learn More
Biomatik Corporation Phospho-STAT5A/B (Tyr694) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51570
Encompass_Preferred
Phospho-STAT5A/B (Tyr694) Colorimetric Cell-Based ELISA Kit for detecting Phospho-STAT5A/B (Tyr694) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42229; P51692. Gene ID: 6776; 6777. Target Synonyms: MGF; MPF; Mammary gland factor; STA5A; STAT5; STAT5A; Signal transducer and activator of transcription 5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1697