Learn More
Biomatik Corporation Phospho-p70 S6 Kinase β (Ser423) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51634
Encompass_Preferred
Phospho-p70 S6 Kinase β (Ser423) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p70 S6 Kinase β (Ser423) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UBS0. Gene ID: 6199. Target Synonyms: S6K2; 70 kDa ribosomal protein S6 kinase 2; p70-S6KB; p70 ribosomal S6 kinase beta; p70 S6Kbeta; p70 S6 kinase beta; S6K-beta; p70-beta; S6 kinase-related kinase; SRK; Serine/threonine-protein kinase 14 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1748