Learn More
Biomatik Corporation Phospho-AML1 (Ser435) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51665
Encompass_Preferred
Phospho-AML1 (Ser435) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AML1 (Ser435) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q01196. Gene ID: 861. Target Synonyms: Acute myeloid leukemia 1 protein; CBF-alpha 2; CBFA2; Core-binding factor; alpha 2 subunit; Oncogene AML-1; PEA2-alpha B; PEBP2-alpha B; Polyomavirus enhancer binding protein 2 alpha B subunit; RUN1; Runt-related transcription factor 1; RUNX1; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1773