Learn More
Biomatik Corporation Phospho-MAPKAPK5 (Thr182) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51678
Encompass_Preferred
Phospho-MAPKAPK5 (Thr182) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAPKAPK5 (Thr182) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8IW41. Gene ID: 8550. Target Synonyms: MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPK5; MAPKAP kinase 5; p38-regulated/activated protein kinase; PRAK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1786