Learn More
Biomatik Corporation Phospho-MAP2K7 (Ser271) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51630
Encompass_Preferred
Phospho-MAP2K7 (Ser271) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAP2K7 (Ser271) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14733. Gene ID: 5609. Target Synonyms: c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; JNK activating kinase 2; JNK kinase 2; JNKK 2; JNKK2; kinase MKK7; MAP kinase kinase 7; MAP2K7; MAPK/ERK kinase 7; MAPKK 7; MP2K7; PRKMK7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9091