Learn More
Biomatik Corporation Phospho-PLK1 (Ser137) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51642
Encompass_Preferred
Phospho-PLK1 (Ser137) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PLK1 (Ser137) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P53350. Gene ID: 5347. Target Synonyms: Polo-like kinase 1; PLK-1; Serine/threonine-protein kinase 13; STPK13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9094