Learn More
Biomatik Corporation Phospho-FRS2 (Tyr436) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51658
Encompass_Preferred
Phospho-FRS2 (Tyr436) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FRS2 (Tyr436) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WU20. Gene ID: 10818. Target Synonyms: FGFR signalling adaptor; FGFR signalling adaptor SNT-1; SNT-1; SNT2; SUC1-associated neurotrophic factor target; Suc1-associated neurotrophic factor target. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9097