Learn More
Biomatik Corporation Phospho-Smad2 (Ser467) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50991
Encompass_Preferred
Phospho-Smad2 (Ser467) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Smad2 (Ser467) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15796. Gene ID: 4087. Target Synonyms: JV18-1; MADH2; MADR2; Mad-related protein 2; Mothers against DPP homolog 2; Mothers against decapentaplegic homolog 2; Smad 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8998