Learn More
Biomatik Corporation Phospho-CD3 ζ (Tyr142) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51096
Encompass_Preferred
Phospho-CD3 ζ (Tyr142) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD3 ζ (Tyr142) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P20963. Gene ID: 919. Target Synonyms: CD3-zeta; T-cell receptor T3 zeta chain; T-cell surface glycoprotein CD3 zeta chain precursor; T3Z; TCRZ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9013