Learn More
Biomatik Corporation Phospho-PP2A-α (Tyr307) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51146
Encompass_Preferred
Phospho-PP2A-α (Tyr307) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PP2A-α (Tyr307) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P67775. Gene ID: 5515. Target Synonyms: PP2A-alpha; PP2AA; RP-C; Replication protein C; Serine/threonine protein phosphatase 2A; catalytic subunit; alpha isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9023