Learn More
Biomatik Corporation Phospho-NF-κB p65 (Ser536) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51523
Encompass_Preferred
Phospho-NF-κB p65 (Ser536) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NF-κB p65 (Ser536) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q04206. Gene ID: 5970. Target Synonyms: NFKB3; nuclear factor NF-kappa-B p65 subunit; p65; RELA; TF65; transcription factor p65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9072