Learn More
Biomatik Corporation Phospho-CD226 (Ser329) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51268
Encompass_Preferred
Phospho-CD226 (Ser329) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD226 (Ser329) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15762. Gene ID: 10666. Target Synonyms: CD226; DNAM1; DNAX accessory molecule-1; PTA1; TLiSA1; adhesion glycoprotein; platelet and T cell activation antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9036