Learn More
Biomatik Corporation Phospho-CXCR4 (Ser339) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51280
Encompass_Preferred
Phospho-CXCR4 (Ser339) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CXCR4 (Ser339) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P61073. Gene ID: 7852. Target Synonyms: C-X-C chemokine receptor type 4; CXC-R4; CXCR-4; Stromal cell-derived factor 1 receptor; SDF-1 receptor; Fusin; Leukocyte-derived seven transmembrane domain receptor; LESTR; LCR1; FB22; NPYRL; HM89; CD184 antigen; CXCR4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9039