Learn More
Biomatik Corporation Phospho-PDGFRβ (Tyr740) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51310
Encompass_Preferred
Phospho-PDGFRβ (Tyr740) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PDGFRβ (Tyr740) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09619. Gene ID: 5159. Target Synonyms: CD140b; kinase PDGFR-beta; PDGF-R-beta; PDGFR; PDGFR-beta; PGFRB; platelet-derived growth factor receptor; beta polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9042