Learn More
Biomatik Corporation Phospho-CRMP2 (Thr514) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51376
Encompass_Preferred
Phospho-CRMP2 (Thr514) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CRMP2 (Thr514) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16555. Gene ID: 1808. Target Synonyms: Collapsin response mediator protein 2; CRMP2; Dihydropyrimidinase related protein-2; DPYL2; DPYSL2; DRP-2; N2A3; TOAD-64; Turned on after division; 64 kDa protein; ULIP 2 protein; ULIP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9048