Learn More
Raybiotech Inc Human Integrin alpha-M/CD11b/MAC-1 ELISA Kit
Supplier:  Raybiotech Inc ELHITGAM1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human Integrin alpha-M.
Catalog No. 50-194-9470