Learn More
Raybiotech Inc Human Kallikrein-13 ELISA Kit
Supplier:  Raybiotech Inc ELHKLK131
Encompass_Preferred
Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Kallikrein-13. Other species not yet determined.
Catalog No. 50-194-9479