Learn More
Raybiotech Inc Human and Mouse Phospho-FRS2 (Tyr436) and Total FRS2 ELISA Kit.
Supplier:  Raybiotech Inc PELFRS2Y436T1
Encompass_Preferred
This assay semi-quantitatively measures FRS2 phosphorylated at Tyrosine-436 as well as total FRS2 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human and Mouse FRS2 phosphorylated at site Tyrosine-436 as well as total FRS2.
Catalog No. 50-195-0600