Learn More
Raybiotech Inc Human Phospho-INSR (Y1189) and Total INSR ELISA Kit.
Supplier:  Raybiotech Inc PELINSRY1189T1
Encompass_Preferred
This assay semi-quantitatively measures phosphorylated INSR (Tyr1189) and Total INSR in lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: The antibody pair provided in this kit recognizes Human INSR phosphorylated at site Tyrosine-1189 and total INSR.
Catalog No. 50-195-0623