Learn More
Raybiotech Inc Human, Mouse Phospho-mTOR (Ser2448) ELISA Kit.
Supplier:  Raybiotech Inc PELMTORS24481
Encompass_Preferred
This assay semi-quantitatively measures mTOR phosphorylated at Serine-2448 in cell lysate samples. RayBio Phosphorylation ELISA kits are very rapid, convenient and sensitive assays that monitor the activation or function of important biological pathways in cell lysate samples. By determining phosphorylated protein in experimental model systems, researchers can verify pathway activation and simultaneously measure numerous different cell lysates without spending excess time and effort in performing Western Blot analyses. Specificity: This ELISA kit recognizes Human, Mouse mTOR phosphorylated at site Serine-2448.
Catalog No. 50-195-0667