Learn More
Biomatik Corporation Human Cementum Protein 1 (CEMP1) ELISA Kit
Supplier:  Biomatik Corporation EKU03070
Encompass_Preferred
Human Cementum Protein 1 (CEMP1) ELISA Kit for detecting CEMP1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6PRD7. Gene ID: 752014. Target Synonyms: CP-23; Cementum Protein-23; Cementoblastoma-derived protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2207