Learn More
Biomatik Corporation Human Galectin 4 (GAL4) ELISA Kit
Supplier:  Biomatik Corporation EKU04322
Encompass_Preferred
Human Galectin 4 (GAL4) ELISA Kit for detecting GAL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56470. Gene ID: 3960. Target Synonyms: LGALS4; Lactose-Binding Lectin 4; L-36 Lactose-Binding Protein; L36LBP; Lectin,Galactoside-Binding Soluble 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2415