Learn More
Biomatik Corporation Rat Laminin (LN) ELISA Kit
Supplier:  Biomatik Corporation EKU05553
Encompass_Preferred
Rat Laminin (LN) ELISA Kit for detecting LN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.0ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E3W9F8; A0A8I5ZQ25; F1MAA7. Gene ID: 298941; 117036. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2603