Learn More
Biomatik Corporation Human Troponin I Type 2, Fast Skeletal (TNNI2) ELISA Kit
Supplier:  Biomatik Corporation EKU07897
Encompass_Preferred
Human Troponin I Type 2, Fast Skeletal (TNNI2) ELISA Kit for detecting TNNI2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48788. Gene ID: 7136. Target Synonyms: AMCD2B; DA2B; FSSV; Troponin I Fast Twitch 2; Troponin I, fast-twitch isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2967