Learn More
Biomatik Corporation Human C-C Motif Chemokine 27 (Ccl27) Elisa Kit
Supplier:  Biomatik Corporation EKC31190
Encompass_Preferred
Human C-C Motif Chemokine 27 (Ccl27) Elisa Kit for detecting C-C Motif Chemokine 27 (Ccl27) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.043ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y4X3; CCL27. Target Synonyms: ALP; C-C motif chemokine 27; CC chemokine ILC; Ccl27; CCL27_HUMAN; Chemokine (C-C motif) ligand 27; CTACK; CTAK; Cutaneous T cell attracting chemokine; Cutaneous T-cell-attracting chemokine; ESkine; IL 11 R alpha locus chemokine; IL-11 R-alpha-locus chemokine; ILC; PESKY; SCYA27; Skinkine; Small inducible cytokine A27; small inducible cytokine subfamily A (Cys-Cys); member 27; Small-inducible cytokine A27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3573