Learn More
Biomatik Corporation Dog Complement 4 (C4) Elisa Kit
Supplier:  Biomatik Corporation EKC32029
Encompass_Preferred
Dog Complement 4 (C4) Elisa Kit for detecting Complement 4(C4) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.8ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3695