Learn More
Biomatik Corporation Human Ficolin 3 (Fcn3) Elisa Kit
Supplier:  Biomatik Corporation EKC33722
Encompass_Preferred
Human Ficolin 3 (Fcn3) Elisa Kit for detecting Ficolin (Collagen/Fibrinogen Domain Containing) 3 (Hakata Antigen) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75636; FCN3. Target Synonyms: FCN3; FCNH; HAKA1; Ficolin-3; Collagen/fibrinogen domain-containing lectin 3 p35; Collagen/fibrinogen domain-containing protein 3; Hakata antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3969