Learn More
Biomatik Corporation Human Protein Gene Product 9.5 (Pgp 9.5) Elisa Kit
Supplier:  Biomatik Corporation EKC35211
Encompass_Preferred
Human Protein Gene Product 9.5 (Pgp 9.5) Elisa Kit for detecting Ubiquitin Carboxyl-Terminal Esterase L1 (Ubiquitin Thiolesterase) in serum, plasma, tissue homogenates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09936; UCHL1. Target Synonyms: Epididymis luminal protein 117; Epididymis secretory protein Li 53; HEL 117; HEL S 53; NDGOA; Neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; Park 5; PARK5; PGP 9.5; PGP9.5; PGP95; Protein gene product 9.5; Ubiquitin C terminal esterase L1; Ubiquitin C terminal hydrolase; Ubiquitin C terminal hydrolase L1; Ubiquitin carboxyl terminal esterase L1; Ubiquitin carboxyl terminal hydrolase isozyme L1; Ubiquitin carboxyl-terminal hydrolase isozyme L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4262