Learn More
Biomatik Corporation Monkey apolipoprotein B100 (Apo-B100) ELISA Kit
Supplier:  Biomatik Corporation EKC36133
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Monkey. Detection Range: 93.75 ng/mL-6000 ng/mL. Sensitivity: 23.44 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q7YQR5. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4349