Learn More
Biomatik Corporation Mouse Neuron-Specific Enolase, Nse Elisa Kit
Supplier:  Biomatik Corporation EKC37479
Encompass_Preferred
Mouse Neuron-Specific Enolase, Nse Elisa Kit for detecting Enolase 2 (Gamma, Neuronal) in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17183; Eno2. Target Synonyms: Eno2; Eno-2Gamma-enolase; EC 4.2.1.11; 2-phospho-D-glycerate hydro-lyase; Enolase 2; Neural enolase; Neuron-specific enolase; NSE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4444