Learn More
Biomatik Corporation Rat Soluble Cluster of differentiation 40 ligand,sCD40L ELISA Kit
Supplier:  Biomatik Corporation EKC39700
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.87 ng/mL-120 ng/mL. Sensitivity: 0.47 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Z2V2. Target Synonyms: RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L, CD40 antigen ligand|T-B cell-activating molecule|TNF-related activation protein|tumor necrosis factor (ligand) superfa. Restriction: For Research Use Only.
Catalog No. 50-229-4643