Learn More
Biomatik Corporation Mouse Interferon Alpha 4 (IFNa4) ELISA Kit
Supplier:  Biomatik Corporation EKU11661
Encompass_Preferred
Mouse Interferon Alpha 4 (IFNa4) ELISA Kit for detecting IFNa4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07351. Gene ID: 15967. Target Synonyms: INFA4; Interferon Alpha-4B; Interferon Alpha-76; Interferon Alpha-M1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3328