Learn More
Biomatik Corporation Rat Creatine Kinase, Mitochondrial 2, Sarcomeric (CKMT2) ELISA Kit
Supplier:  Biomatik Corporation EKU12196
Encompass_Preferred
Rat Creatine Kinase, Mitochondrial 2, Sarcomeric (CKMT2) ELISA Kit for detecting CKMT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09605. Target Synonyms: MtCK; CK-MT2; SMTCK; Mib-CK; Creatine Kinase S-type; Basic-type mitochondrial creatine kinase; Sarcomeric mitochondrial creatine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3450