Learn More
Biomatik Corporation Green Fluorescent Protein (GFP) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12468
Encompass_Preferred
Green Fluorescent Protein (GFP) Anti-antigen ELISA Kit for detecting GFP in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P42212. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3528